Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Carfentanil-13C6 oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001141 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-141 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-141 Supplier MilliporeSigma Supelco Supplier No. C2031EA

missing translation for 'validMainframeMsg'

Certified reference material

This stable labeled internal standard is suitable for quantitation of carfentanil in fentanyl GC/MS or LC/MS methods for applications in clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution. Marketed under the trade name Wildnil as a general anesthetic for large animals, carfentanil hit the illicit market in 2016 as a new NPS drug or in combination with heroin. Carfentanil, also known as 4-carbomethoxyfentanyl, is a fentanyl analog 10,000 times more potent than morphine, making it one of the most potent synthetic opioids available. This product will be shipped in a can containing one ampoule of C-203-0.5ML due to the compound's acute toxicity.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula 13C6C18H30N2O3 · C2H2O4
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 490.50
Packaging Ampule
Quantity 0.5 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.