Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Carisoprodol-13C3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1341ML

Catalog No. 01-001-095


Add to Cart

Description

Description

This stable-labeled internal standard is suitable for carisoprodol testing or isotope dilution methods by LC/MS or GC/MS for applications in forensic analysis, urine drug testing, clinical toxicology, or pain prescription monitoring. Sold under trade names such as Soma and Sanoma, carisoprodol is a skeletal muscle relaxant that is often abused recreationally. Carisoprodol is purportedly used by drug users to potentiate the effects of other narcotics.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
263.31
1 mL
C913C3H24N2O4
100 μg/mL in methanol
Ampule
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation