Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Catecholamine Metabolites Mix Solution, 1 mg/mL each component in methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258416 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-416 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-416 Supplier MilliporeSigma Supelco Supplier No. C1111ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Certified Spiking Solution™ of major catecholamine metabolites 5-Hydroxyindole-3-acetic acid (5-HIAA), Homovanillic acid (HVA), and (±)-Vanilmandelic acid (VMA).

This multi-component mix is suitable for use in preparation of linearity standards, calibrators, or controls as well as for other critical quantitative applications such as internal proficiency testing or for OEM use in preparation of reagents, assay development, and instrument IQ/OQ.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Molecular Formula C9H10O5
Concentration 1 mg/mL each component in Methanol
Industry Type Clinical Testing, Forensics, Toxicology
Molecular Weight (g/mol) 198.17
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.