Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cetirizine DiHydrochloride Solution, 1.0 mg/mL in Acetonitrile:Water (9:1), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1581ML

Catalog No. 01-001-114


Add to Cart

Description

Description

Cetirizine is a major metabolite of the first generation antihistamine hydroxyzine. Cetirizine is also a second generation antihistamine that is sold under the trade name Zyrtec™. This Certified Spiking Solution™ is suitable for use in a variety of GC/MS or LC/MS testing applications in forensic toxicology, urine drug testing, or clinical chemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 83881-52-1
MFCD00941428
1.0 mg/mL in acetonitrile:water (9:1) (as free base)
Ampule
2°C (35.6°F) - closed cup
C21H25ClN2O3 · 2HCl
UN 1648 3 / PGII
461.81
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation