Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Chlordiazepoxide-d5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001150 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-150 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-150 Supplier MilliporeSigma Supelco Supplier No. C9121ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Chlordiazepoxide is a benzodiazepine sold under trade names such as Librax™ or Klopoxid for short-term treatment of anxiety or management of acute alcohol withdrawal syndrome. This stable-labeled internal standard is suitable for chlordiazepoxide testing or isotope dilution methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 65891-81-8
Linear Formula C16D5H9ClN3O
MDL Number MFCD00057322
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 304.79
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.