Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-cis-11-Nor-9-carboxy-Delta9-THC-D3 glucuronide Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002077 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-077 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-077 Supplier MilliporeSigma Supelco Supplier No. T0801ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A stable-labeled internal standard for 11-Nor-9-carboxy-δ9-THC glucuronide, a major urinary metabolite of δ9-THC. This Certified Spiking Solution™ is suitable for LC/MS or GC/MS applications such as forensic testing, toxicology analysis, urine drug testing, or isotope dilution methods. This internal standard provides added value in allowing for quantitation of glucuronide levels by LC-MS/MS without need of an added hydrolysis step.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C27H33O10D3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Synonym (+/-)-cis-11-Nor-9-carboxyDelta9-THC-d3 glucuronide
Formula Weight 523.59
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.