Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Clomipramine-d3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001079 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-079 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-079 Supplier MilliporeSigma Supelco Supplier No. C1161ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Clomipramine is a tricyclic antidepressant used to many conditions from major depression and panic disorder to narcolepsy and obsessive compulsion disorder (OCD). This internal standard is is suitable for quantitation of Clomipramine levels in urine, serum, or plasma by LC/MS or GC/MS for applications in clinical toxicology, forensic analysis, isotope dilution methods, or pharmaceutical research.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1398065-86-5
Linear Formula C19D3H20ClN2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Clomipramine-D3 hydrochloride solution
Formula Weight 354.33
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.