Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Clonazepam-d4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C9051ML

Catalog No. 01-001-146


Add to Cart

Description

Description

Clonazepam, sold under trade names Klonopin™ or Rivotril, is a long-acting, potent benzodiazepine approved for the treatment of epilepsy. This stable-labeled internal standard is suitable for use in LC/MS or GC/MS applications including forensic analysis, clinical toxicology, and urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 170082-15-2
MFCD00798470
100 μg/mL in methanol
319.74
1 mL
C15D4H6ClN3O3
UN 1230 - class 3 - PG 2 - Methanol, solution
Clonazepam-d4 solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation