Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Clonazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001148 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-148 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-148 Supplier MilliporeSigma Supelco Supplier No. C9071ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Clonazepam, marketed under trade names such as Klonopin and Rivotril, is a potent, long-acting benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant and hypnotic properties. Clonazepam is prescribed for the treatment of epilepsy. This certified Snap-N-Spike™ solution is suitable for use in GC/MS or LC/MS applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1622-61-3
Linear Formula C15H10ClN3O3
MDL Number MFCD00057746
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 315.71
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.