Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Clozapine Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258367 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-367 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-367 Supplier MilliporeSigma Supelco Supplier No. C0591ML
Add to Cart
Add to Cart

A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC/MS or GC/MS.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
CAS 5786-21-0
Product Line CERILLIANT
Physical Form Liquid
Molecular Formula C18H19ClN4
Concentration 1 mg/mL in Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 326.82
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.