Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cocaethylene Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258362 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-362 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-362 Supplier MilliporeSigma Supelco Supplier No. C0101ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for GC/MS or LC/MS methods in clinical toxicology, forensic analysis, or urine drug testing.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
CAS 529-38-4
Product Line CERILLIANT
Physical Form Liquid
Molecular Formula C18H23NO4
Concentration 1 mg/mL in Acetonitrile
MDL Number MFCD00871900
Synonym Benzoylecgonine ethyl ester
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 317.38
Quantity 1 mL
Grade Certified Reference Material
Show More Show Less

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.