Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Corticosterone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1171ML

Catalog No. 01-001-080


Add to Cart

Description

Description

Corticosterone is a corticosteroid and aldosterone precursor produced in the adrenal glands. Serum corticosterone levels are measured by LC-MS/MS for newborn screening and diagnosis of 11-hydroxylase defficiency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 50-22-6
MFCD00037715
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C21H30O4
UN 1230 - class 3 - PG 2 - Methanol, solution
346.46
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation