Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cyanocobalamin (Vitamin B12) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002144 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-144 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-144 Supplier MilliporeSigma Supelco Supplier No. V0191ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Cyanocobalamin is the most common and widely-produced of the vitamin B12 analogs. Vitamin B12, important for proper neuronal function and formation of blood cellular components, is monitored directly in patient serum samples by LC-MS/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 68-19-9
Linear Formula C63H88CoN14O14P
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym Vitamin B12, Vitamin B12, alpha-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin
Formula Weight 1355.37
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.