Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cyclobenzaprine-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001077 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-077 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-077 Supplier MilliporeSigma Supelco Supplier No. C1141ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Cyclobenzaprine is a muscle relaxant sold under the trade name Flexeril™ used to relieve skeletal muscle spasms and associated pain from acute musculoskeletal conditions. This internal standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing, pain prescription monitoring and isotope dilution methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C20H18ND3HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Cyclobenzaprine-D3 hydrochloride
Formula Weight 314.87
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.