Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cyclosporin D Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1081ML

Catalog No. 01-001-074


 Restricted Item
Add to Cart

Description

Description

An internal standard for clinical testing of Cyclosporin A by LC-MS/MS. This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within Cyclosporin A's narrow therapeutic range. The Snap-N-Spike™ format allows laboratories to eliminate weighing operations of these hazardous substances, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 63775-96-2
UN 1648 3 / PGII
1-(3-Chlorophenyl)piperazine-D8 hydrochloride solution
1 mL
MFCD11111526
1.0 mg/mL in acetonitrile
Ampule
2°C (35.6°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

EINECSNumber : 200-835-2

Recommended Storage : -70°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation