Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dehydro Aripiprazole-D8 Solution, 100 μg/mL (Methanol with 5% 1 N HCl), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001253 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-253 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-253 Supplier MilliporeSigma Supelco Supplier No. D1781ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for use in LC/MS or GC/MS applications including forensic or clinical toxicology analysis and urine drug testing. Dehydro Aripiprazole is the primary, active metabolite of aripiprazole, marketed as the atypical antipsychotic Abilify™ for the treatment of schizophrenia, bipolar disorder, and clinical depression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C23H17D8Cl2N3O2
Concentration 100 μg/mL (methanol with 5% 1 N HCl)
Formula Weight 454.42
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.