Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001186 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-186 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-186 Supplier MilliporeSigma Supelco Supplier No. D0651ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

DHEAS is a metabolite of dehydroepiandrosterone (DHEA), an endogenous steroid that has been implicated in a broad range of biological effects in humans and other mammals. DHEAS levels are measured by LC-MS/MS for determination of excess adrenal activity as observed in adrenal cancer, certain forms of congenital adrenal hyperplasia (CAH), and polycystic ovary syndrome.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1099-87-2
Linear Formula C19H28NaO5S
MDL Number MFCD00010470
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free sulfate)
Synonym Dehydroepiandrosterone 3-sulfate sodium salt
Formula Weight 391.48
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.