Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (-)-Delta8-THC Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0321ML

Catalog No. 01-002-046


Add to Cart

Description

Description

A certified solution standard suitable for LC/MS or GC/MS applications in cannabinoid testing, clinical toxicology, forensic analysis, or urine drug testing. δ8-THC is an impurity in synthetic THC manufacturing processes. This cannabinoid has antiemetic, anxiolytic, appetite-stimulating, and analgesic properties.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 5957-75-5
UN 1230 - class 3 - PG 2 - Methanol, solution
314.46
1 mL
C21H30O2
1.0 mg/mL in methanol
Ampule
11°C (51.8°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation