Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Desalkylflurazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D9151ML

Catalog No. 01-001-266


 Restricted Item
Add to Cart

Description

Description

Desalkylflurazepam, also known as N-desalkyl-2-oxoquazepam, is a long-acting metabolite of the benzodiazepine drug flurazepam. Flurazepam is sold unde the trade names Dalmane and Dalmadorm for the treatment of mild to moderate insomnia. This Certified Spiking Solution™ is suited for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 2886-65-9
UN 1230 - class 3 - PG 2 - Methanol, solution
288.70
1 mL
C15H10ClFN2O
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation