Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Desmethylolanzapine DiHydrochloride Solution, 1.0 mg/mL in Acetonitrile:Water (1:1), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0691ML

Catalog No. 01-001-189


Add to Cart

Description

Description

Desmethylolanzapine is the primary metabolite of olanzapine, marketed as the atypical antipsychotic Zyprexa™for the treatment of schizophrenia and bipolar disorder. This new certified solution standard is suitable for use in LC or GC applications including clinical toxicology analysis, forensic testing, or therapeutic drug monitoring.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 161696-76-0
MFCD29917817
1.0 mg/mL in acetonitrile:water (1:1) (as free base)
371.33
1 mL
C16H18N4S · H2Cl2
UN 1648 3 / PGII
Desmethylolanzapine dihydrochloride solution
Ampule
6°C (42.8°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation