Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Desomorphine Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001203 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-203 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-203 Supplier MilliporeSigma Supelco Supplier No. D0831ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Desomorphine, most commonly referred to by its street name Krokodil, is a designer opiate and morphine derivative originally abused in Russia and other parts of Europe due to its ease of manufacture from codeine. Desomophine was previously sold under the trade name Permonid as an opiate drug for treatment of pain. This certified solution standard is suitable for use in LC/MS or GC/MS opiate testing applications in clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 427-00-9
Linear Formula C17H21NO2
MDL Number MFCD00864242
UN Number UN 1648 3 / PGII
Concentration 1 mg/mL in acetonitrile
Formula Weight 271.35
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.