Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dextromethorphan-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0711ML

Catalog No. 01-001-191


Add to Cart

Description

Description

Dextromethorphan, also known as DXM or DM, is an opiate cough suppressant also used recreationally as an illicit drug and hallucinogen. This stable labeled internal standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology, urine drug testing, and forensic analysis to pain management testing and isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 524713-56-2
UN 1230 - class 3 - PG 2 - Methanol, solution
274.42
1 mL
C18H22D3NO
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation