Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dextrorphan tartrate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0341ML

Catalog No. 01-001-165


Add to Cart

Description

Description

A Snap-N-Spike™ Reference Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, P450 trials, or pharmaceutical research. Dextrorphan is a primary metabolite of dextromethorphan in blood. Dextromethorphan, an active ingredient in many over-the-counter cold and cough medicines, is used recreationally as a hallucinogen with effects similar to ketamine and phencyclidine (PCP).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 143-98-6
MFCD00151495
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C21H29NO7
UN 1230 - class 3 - PG 2 - Methanol, solution
407.46
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation