Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Diazepam-d5 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D9101ML

Catalog No. 01-001-265


Add to Cart

Description

Description

Diazepam is a benzodiazepine used to treat a number of symptoms from anxiety and panic attacks to insomnia. This stable-labeled internal standard is suited for quantitation of diazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Diazepam, a benzodiazpine frequently abused recreationally, is sold under the trade name Valium™.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 65854-76-4
MFCD00070151
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C16D5H8ClN2O
UN 1230 - class 3 - PG 2 - Methanol, solution
289.77
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation