Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dibutylone Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001240 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-240 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-240 Supplier MilliporeSigma Supelco Supplier No. D1611ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Dibutylone is a synthetic stimulant of the amphetamine and cathinone classes also known as bk-DMBDB. Significant abuse of this designer drug has been reported in Europe and the US. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for GC/MS or LC/MS testing applications in forensic analysis, urine drug testing or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 17763-12-1
Linear Formula C13H17NO3 HCl
MDL Number MFCD27966866
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 271.74
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.