Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Diclazepam Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001238 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-238 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-238 Supplier MilliporeSigma Supelco Supplier No. D1591ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Diclazepam, also known as, chlorodiazepam and 2′-chloro-diazepam, is a benzodiazepine and functional analog of diazepam that has emerged on the designer drug market. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for use in LC/MS or GC/MS benzodiazepine testing methods in clinical toxicology, urine drug testing, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 2894-68-0
Linear Formula C16H12Cl2N2O
MDL Number MFCD01237640
UN Number UN 1648 3 / PGII
Concentration 1 mg/mL in acetonitrile
Formula Weight 319.19
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.