Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dihydrocodeine-6ß-D-glucuronide Solution, 1.0 mg/mL in Methanol:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001221 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-221 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-221 Supplier MilliporeSigma Supelco Supplier No. D1091ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Dihydrocodeine-6β-D-glucuronide is a major urinary metabolite of dihydrocodeine, an opiate used for treatment of moderate to severe pain and as an antitussive. Dihydrocodeine is twice as potent as codeine and is frequently abused in Europe and in Asia. This Certified Spiking Solution™ is suitable for use as a starting material in calibrators and controls for LC/MS and GC/MS testing methods in clinical toxicology, forensic analysis, pharmaceutical research, and pain prescription monitoring.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 88480-40-4
Linear Formula C24H31NO9
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol:water (1:1)
Formula Weight 477.50
Packaging Ampule
Quantity 1 mL
Flash Point 24°C (75.2°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.