Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dihydrocodeine-D6 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001207 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-207 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-207 Supplier MilliporeSigma Supelco Supplier No. D0871ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Dihydrocodeine, sold under brand names such as Drocode, Paracodin, Hydrocodin, and Fortuss, is a semi-synthetic opiate and opioid analgesic prescribed for pain, severe dyspnea, and as an antitussive. This internal standard is suitable for dihydrocodeine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, pain prescription monitoring, or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C18H17D6NO3·HCl
MDL Number MFCD01861525
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 343.88
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.