Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Dihydromorphine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Dihydromorphine is a semisynthetic opioid analgesic sold under the trade names Paramorfan or Paramorphan for treatment of pain. Dihydromorphine is also a primary active metabolite of the analgesic opioid drug dihydrocodeine. This Snap-N-Spike™ Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing and pain prescription monitoring to pharmaceutical research.
Specifications
Specifications
| CAS | 67-56-1, 509-60-4 |
| Linear Formula | C17H21NO3 |
| MDL Number | MFCD00190622 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol |
| Formula Weight | 287.35 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?