Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Diltiazem Hydrochloride Solution, 1.0 mg/mL in Acetonitrile (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0351ML

Catalog No. 01-001-166


Add to Cart

Description

Description

A Certified Spiking Solution™ produced for use in clinical toxicology, forensic analysis, or pharmaceutical research applications by GC/Ms or LC/MS. Diltiazem is a pharmaceutical drug in the calcium channel blocker class. Diltiazem is prescribed for the treatment of hypertension and heart conditions such as angina pectoris and some types of arrhythmia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 33286-22-5
MFCD00069252
1.0 mg/mL in acetonitrile (as free base)
Ampule
2°C (35.6°F) - closed cup
C22H27ClN2O4S
UN 1648 3 / PGII
450.98
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

EINECSNumber : 200-835-2

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation