Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Diphenhydramine-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0171ML

Catalog No. 01-001-159


Add to Cart

Description

Description

A stable-labeled internal standard for diphenhydramine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, or forensic analysis. Diphenhydramine is an antihistamine sold over the counter under the trade name Benadryl™. Dihydramine is also consumed recreationally as a euphoriant and a deliriant.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 170082-18-5
MFCD01861524
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C17D3H18NO
UN 1230 - class 3 - PG 2 - Methanol, solution
258.37
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation