Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Diphenhydramine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001158 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-158 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-158 Supplier MilliporeSigma Supelco Supplier No. D0151ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Diphenhydramine is an antihistamine sold over the counter under the trade name Benadryl™ and in other over the counter medications such as Tylenol PM, Advil PM, and Excedrin PM. Dihydramine is also consumed recreationally as a euphoriant and a deliriant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 147-24-0
Linear Formula C17H22ClNO
MDL Number MFCD00012479
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 291.82
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.