Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Dopamine Hydrochloride Solution, 1.0 mg/mL in Methanol with 5% 1 M HCl (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001201 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-201 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-201 Supplier MilliporeSigma Supelco Supplier No. D0811ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Dopamine is a catecholamine which acts as a neurotransmitter for dopamine receptors and neurohormone for regulation of numerous brain functions. This certified solution and linearity standard is suitable for LC/MS applications in diagnostic testing, endocrinology, and clinical chemistry.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 62-31-7
Linear Formula C8H11NO2·HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol with 5% 1 M HCl (as free base)
Formula Weight 189.64
Packaging Ampule
Quantity 1 mL
Flash Point 11°C (51.8°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.