Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Doxepin Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D9271ML

Catalog No. 01-001-272


Add to Cart

Description

Description

Doxepin is a tricyclic antidepressant used for the treatment of sleep maintenance and sold under trade names such as Silenor™ and Aponal. This analytical reference standard is appropriate for use as starting material in calibrators or controls for numerous LC/MS or GC/MS applications such as forensic analysis, clinical toxicology, urine drug testing, or pharmaceutical research.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1229-29-4
UN 1230 - class 3 - PG 2 - Methanol, solution
315.84
1 mL
C19H22ClNO
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation