Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Duloxetine-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0681ML

Catalog No. 01-001-188


Add to Cart

Description

Description

A stable-labeled internal standard suitable for numerous GC/MS and LC/MS applications from clinical toxicology and urine drug testing to forensic analysis and isotope dilution methods. Duloxetine, marketed in the US under the trade name Cymbalta™, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD09752117
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C18D3H16NOS · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
336.89
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation