Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Duloxetine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001169 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-169 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-169 Supplier MilliporeSigma Supelco Supplier No. D0441ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Duloxetine, marketed in the US under the trade name Cymbalta™, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis. This Certified Spiking Solution™ is applicable for use in applications such as clinical toxicology, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 136434-34-9
Linear Formula C18H20ClNOS
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym Duloxetine hydrochloride solution
Formula Weight 333.88
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.