Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Ecgonine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001275 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-275 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-275 Supplier MilliporeSigma Supelco Supplier No. E0041ML

missing translation for 'validMainframeMsg'

Certified reference material

Ecgonine is not only a precursor of cocaine, but also a metabolite obtained by cocaine hydrolysis. This analytical reference standard is suitable for use as starting material in calibrators and controls for a variety of LC/MS or GC/MS applications in cocaine testing from forensic analysis and clinical toxicology to urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 5796-31-6
Linear Formula C9H15NO3·HCl
MDL Number MFCD00069267
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 221.68
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.