Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Ecgonine methyl ester-D3 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E0021ML

Catalog No. 01-001-274


Add to Cart

Description

Description

Ecgonine methyl ester is a major urinary metabolite of cocaine, a popular illicit drug well known as 'crack' or the free base form of cocaine that can be smoked. This stable-labeled internal standard is suitable for use in cocaine testing or isotope dilution methods for GC/MS or LC/MS applications in urine drug testing, clinical toxicology or forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 136765-34-9
UN 1648 3 / PGII
Ecgonine methyl ester-d3
Ampule
2°C (35.6°F) - closed cup
C10H14D3NO3
100 μg/mL in acetonitrile
202.27
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation