Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ EDDP-D3 perchlorate Solution, 1.0 mg/mL in Methanol (as pyrrolinium), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001292 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-292 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-292 Supplier MilliporeSigma Supelco Supplier No. E0621ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

EDDP is the major urinary metabolite of methadone, a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. This stable-labeled internal standard is suitable for EDDP and methadone testing methods by GC/MS or LC/MS for forensic analysis, urine drug testing or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 136765-23-6
Linear Formula C20D3H21N · ClO4
MDL Number MFCD00673273
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as pyrrolinium)
Synonym EDDP-D3 perchlorate solution
Formula Weight 380.88
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.