Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ EDDP perchlorate, , Sold by MilliporeSigma™ Supelco™

Catalog No. 01001276 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-276 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-276 Supplier MilliporeSigma Supelco Supplier No. E00610MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Reference Material of the major urinary metabolite of methadone. Methadone, sold under various trade names such as Symoron, Amidone or Methadose™, is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. Suitable uses for this reference standard include as starting material in preparation of calibrators and controls for LC/MS or GC/MS methods in methadone testing for applications in clinical toxicology or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 31161-17-8
Linear Formula C20H23N · HClO4
UN Number UN 2811 6.1 / PGIII
Synonym EDDP perchlorate, 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine perchlorate
Formula Weight 377.86
Packaging Ampule
Quantity 10 mg

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.