Learn More
Sigma Organic Chemistry Cerilliant (±)-Epinephrine-D6 Solution, Certified Reference Material, 100 µg/mL, 1 mL
Catalog No. NC4042888 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
(±)-Epinephrine-D6 solution is a certified reference material used as an internal standard for the quantification of catecholamine levels in clinical samples. It is suitable for diagnostic testing in endocrinology, specifically for monitoring markers associated with pheochromocytoma, paraganglioma, and neuroblastoma via LC-MS/MS or GC-MS techniques.
  • Provided as a 100 μg/mL single component solution in methanol with 5% 1 M HCl.
  • Supplied in a 1 mL ampule for convenient use.
  • Features Snap-N-Spike technology for reliable analytical performance.
  • Suitable for both liquid chromatography and gas chromatography applications.
  • Requires storage at −70°C in an ultra freezer.
Catalog No. NC4042888 Supplier Sigma Organic Chemistry Supplier No. E0771ML
  • NEW LOWER PRICE
(±)-Epinephrine-D6 solution is a certified reference material used as an internal standard for the quantification of catecholamine levels in clinical samples. It is suitable for diagnostic testing in endocrinology, specifically for monitoring markers associated with pheochromocytoma, paraganglioma, and neuroblastoma via LC-MS/MS or GC-MS techniques.
  • Provided as a 100 μg/mL single component solution in methanol with 5% 1 M HCl.
  • Supplied in a 1 mL ampule for convenient use.
  • Features Snap-N-Spike technology for reliable analytical performance.
  • Suitable for both liquid chromatography and gas chromatography applications.
  • Requires storage at −70°C in an ultra freezer.