Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Cerilliant (±)-Epinephrine Hydrochloride Solution, Certified Reference Material, 1 mg/mL, 1 mL

Catalog No. NC4042885 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

(±)-Epinephrine hydrochloride is a certified reference material provided as a 1 mg/mL solution in methanol. It is designed for use as a catecholamine marker in diagnostic testing and endocrinology, specifically for LC-MS/MS monitoring of epinephrine in urine or plasma samples.

  • Concentration of 1 mg/mL (as free base)
  • Supplied in a 1 mL ampule
  • Suitable for LC-MS/MS diagnostic applications
  • Adrenergic receptor agonist biological activity
  • Storage temperature between -10 and -25 °C
  • Certified reference material grade

Catalog No. NC4042885 Supplier Sigma Organic Chemistry Supplier No. E1431ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Epinephrine hydrochloride is a certified reference material provided as a 1 mg/mL solution in methanol. It is designed for use as a catecholamine marker in diagnostic testing and endocrinology, specifically for LC-MS/MS monitoring of epinephrine in urine or plasma samples.

  • Concentration of 1 mg/mL (as free base)
  • Supplied in a 1 mL ampule
  • Suitable for LC-MS/MS diagnostic applications
  • Adrenergic receptor agonist biological activity
  • Storage temperature between -10 and -25 °C
  • Certified reference material grade

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.