Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Estriol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001301 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-301 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-301 Supplier MilliporeSigma Supelco Supplier No. E0741ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Estriol is a metabolite of estradiol and the primary estrogen found in urine. Estriol levels in serum, plasma, or urine are measured by LC-MS/MS during pregnancy as part of maternal screens for Down-Syndrome-afflicted fetuses.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 50-27-1
Linear Formula C18H24O3
MDL Number MFCD00003691
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 288.38
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.