Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Ethanol-40, 40 mg/dL in H2O, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258383 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-383 10 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-383 Supplier MilliporeSigma Supelco Supplier No. E04510X1.2ML
Add to Cart
Add to Cart

A kit of NIST traceable ethanol standards suitable for calibration of ethanol testing methods. These Certified Reference Materials are widely used in forensic analysis and clinical toxicology applications for determination of blood alcohol content.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-17-5
Quantity 10 x 1.2 mL
Physical Form Liquid
Molecular Formula C2H5OH
MDL Number MFCD00003568
Beilstein 1718733
Synonym Ethanol solution, Ethyl alcohol
Molecular Weight (g/mol) 46.07
Concentration or Composition (by Analyte or Components) 40 mg/dL in H2O

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.