Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Etizolam-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001307 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-307 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-307 Supplier MilliporeSigma Supelco Supplier No. E0821ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for quantitation of etizolam levels in urine or other matrices by LC/MS or GC/MS in forensic analysis, clinical toxicology, or urine drug testing applications. Etizolam, sold under brand names such as Etilaam, Etizola, or Depas, is a benzodiazepine analog used for the short-term treatment of insomnia, anxiety, or panic attacks. Etizolam is also sold on the designer drug market with cases of abuse noted in Europe and the US.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C17H12D3ClN4S
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 345.86
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.