Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Eutylone-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001314 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-314 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-314 Supplier MilliporeSigma Supelco Supplier No. E0911ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Eutylone is a synthetic cathinone found in numerous bath salt blends under the street name bk-MBDB. This designer drug and stimulant is abused in the US and Europe. This stable-labeled internal standard is suitable for quantification of eutylone using LC/MS or GC/MS methods for applications in clinical toxicology, forensic analysis, and urine drug testing applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C13H12D5NO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 276.77
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.