Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Fenproporex-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001380 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-380 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-380 Supplier MilliporeSigma Supelco Supplier No. F0571ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable isotope labeled Snap-N-Shoot™ Reference Solution is suitable for quantification of fenproporex in a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing and to clinical toxicology and sports testing. Fenproporex, also known as Perphoxene and used as an appetite suppressant for the treatment of obesity, is a stimulant drug of the phenethylamine and amphetamine classes that produces amphetamine as a metabolite. Fenproporex is designated a Schedule IV controlled substance in the US pursuant to the Controlled Substances Act and is also listed as banned substance by the World Anti-Doping Agency.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C12H11D5N2 · HCl
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 229.76
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.