Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Flecainide Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco F0171ML

Catalog No. 01-001-352


Add to Cart

Description

Description

Flecainide, marketed under trade names such as Almarytm, Apocard, Ecrinal™, and Flécaine, is a class Ic antiarrhythmic drug used to prevent and treat a variety of cardiac arrhythmias. This Certified Spiking Solution™ is suitable for use as a starting material in the preparation of calibrators, controls, or linearity standards in flecainide therapeutic drug monitoring methods by LC/MS or LC that ensure patients remain within the drug's narrow therapeutic range.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 54143-55-4
MFCD00864713
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C17H20F6N2O3
UN 1230 - class 3 - PG 2 - Methanol, solution
414.34
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation