Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Flunitrazepam-d7, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco F9151ML

Catalog No. 01-001-396


Add to Cart

Description

Description

A stable-labeled internal standard of flunitrazepam suitable for quantitation of flunitrazepam levels in LC/MS or GC/MS applications for clinical toxicology, urine drug testing, and forensic analysis. Marketed under the trade name Rohypnol, flunitrazepam is an intermediate-acting benzodiazepine typically prescribed for the treatment of severe insomnia. Flunitrazepam is noted for its sedative, muscle relaxant, anxiolytic, and anticonvulsant properties.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1286448-08-5
UN 1230 - class 3 - PG 2 - Methanol, solution
Flunitrazepam-d7 solution
Ampule
9.7°C (49.5°F) - closed cup
C16D7H5FN3O3
100 μg/mL in methanol
320.33
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation