Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Fluoxetine-D6, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco F9191ML

Catalog No. 01-001-398


Add to Cart

Description

Description

A Certified Spiking Solution™ suitable as an internal standard for quantitation of Fluoxetine levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. Fluoxetine is a pharmaceutical antidepressant which functions as a selective serotonin reuptake inhibitor (SSRI). Fluoxetine is sold under the trade name Prozac for treatment of psychological conditions including major depression and obsessive-compulsive disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD08276449
100 μg/mL in methanol (as free base)
405.40
1 mL
C17D6H12F3NO · C2H2O4
UN 1230 - class 3 - PG 2 - Methanol, solution
Fluoxetine-D6 oxalate solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation